Isorhapontigenin ≥96.0% (by GC)
Supplier: TCI America
Synonyms:
(E)-5-(4-hydroxy-3-methoxystyryl)benzene-1,3-diol, 3,4',5-Trihydroxy-3'-methoxy-trans-stilbene
I0804-100MG
TCI0804-100MGEA
107.8
USD
TCI0804-100MG
Isorhapontigenin ≥96.0% (by GC)
Isorhapontigenin
CAS Number: 32507-66-7
MDL Number: MFCD12407151
Molecular Formula: C15H14O4
Molecular Weight: 258.27
Purity/Analysis Method:
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antioxidants, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 3,4',5-Trihydroxy-3'-methoxy-trans-stilbene
Formula:
C₁₅H₁₄O₄ MW: 258.27 g/mol Melting Pt: 182 °C |
MDL Number:
MFCD12407151 CAS Number: 32507-66-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...