Iron(II) acetate ≥90.0%
Supplier: TCI America
I0765-25G
I0765-5G
TCI0765-25GEA
155.58
USD
TCI0765-25G
TCI0765-5G
Iron(II) acetate ≥90.0%
Iron (II) acetate
CAS Number: 3094-87-9
MDL Number: MFCD00058909
Molecular Formula: C4H6O4Fe
Molecular Weight: 173.93
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction
Other Product Names / Synonyms: Acetic Acid Iron(II) Salt
Formula:
C₄H₆FeO₄ |
MDL Number:
MFCD00058909 CAS Number: 3094-87-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...