Iodoacetamide ≥98.0% (by total nitrogen basis) for biochemical research

Supplier: TCI America

I0741-5G
TCI0741-5GEA 30.09 USD
TCI0741-5G
Iodoacetamide ≥98.0% (by total nitrogen basis) for biochemical research
Iodoacetamide

[for Biochemical Research]
CAS Number: 144-48-9
MDL Number: MFCD00008028
Molecular Formula: C2H4INO
Molecular Weight: 184.96
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 95

Formula: ICH₂CONH₂
Melting Pt: 95 °C
MDL Number: MFCD00008028
CAS Number: 144-48-9
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49c82c20ad9817712cc47f0df9beae9c15f15dd1&vid=f6dd1df254ec17adf852f8d04a695a4bcd50453d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR