Isosorbide dimethyl ether ≥98.0%

Supplier: TCI America

Synonyms: (3R,3aR,6S,6aR)-3,6-Dimethoxyhexahydrofuro[3,2-b]furan

I0390-25G
TCI0390-025GEA 53.4 USD
TCI0390-025G
Isosorbide dimethyl ether ≥98.0%
Isosorbide dimethyl ether

CAS Number: 5306-85-4 MDL Number: MFCD00064828 Molecular Formula: C8H14O4 Molecular Weight: 174.20 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Flash Point (°C): 108 Specific Gravity (20/20): 1.17

Formula: C₈H₁₄O₄
Density: 1.17 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00064828
CAS Number: 5306-85-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c4a07c871f692856437dec36f85de3ea1b6366c&vid=4c4a07c871f692856437dec36f85de3ea1b6366c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR