Ethyl-4-iodobenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Ethyl 4-iodobenzoate, 4-Iodobenzoic acid ethyl ester

I0369-25G I0369-5G
TCI0369-025GEA 51 USD
TCI0369-025G TCI0369-5G
Ethyl-4-iodobenzoate ≥98.0% (by GC)
Ethyl 4-iodobenzoate

CAS Number: 51934-41-9 MDL Number: MFCD00017344 Molecular Formula: C9H9IO2 Molecular Weight: 276.07 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 283 Flash Point (°C): 110 Specific Gravity (20/20): 1.66


Other Product Names / Synonyms: 4-Iodobenzoic Acid Ethyl Ester

Formula: C₉H₉IO₂
Boiling Pt: 283 °C
Density: 1.66 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00017344
CAS Number: 51934-41-9
EINECS: 429-550-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cba82d0b4d52c22ae902141ac3a49b0b91a511a6&vid=cf9104727a87f0fe3bdd4c65befd917e9e46b494&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR