Tetrabutylammonium dihydrogen phosphate 0.5 mol/L in water

Supplier: TCI America

Synonyms: Tetra-n-butylammonium phosphate monobasic, Tetrabutylammonium phosphate monobasic, Tetra-n-butylammonium dihydrogen phosphate, IPC-TBA-P, Tetra-n-butylammonium phosphate

I0367-10ML I0367-100ML
TCI0367-010MLEA 23 USD
TCI0367-010ML TCI0367-100ML
Tetrabutylammonium dihydrogen phosphate 0.5 mol/L in water
Tetrabutylammonium dihydrogen phosphate

[Reagent for Ion-Pair Chromatography]
CAS Number: 5574-97-0
MDL Number: MFCD00064526
Molecular Formula: C16H37NO
Form: Clear Liquid
Color: Colorless


Other Product Names / Synonyms: IPC-TBA-P

Formula: C₁₆H₃₈NO₄P
MW: 339.46 g/mol
MDL Number: MFCD00064526
CAS Number: 5574-97-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f6d560c8aa2c3d4f50e191c939ef6914d5956c6&vid=59ef918ee2a231ffcbb75867090cc3b57f6180d3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR