Tetraethylammonium hydroxide 10% in water
Supplier: TCI America
Synonyms:
Tetrylammonium hydroxide
I0363-25ML
TCI0363-25MLEA
62
USD
TCI0363-25ML
Tetraethylammonium hydroxide 10% in water
Tetraethylammonium hydroxide
[Reagent for Ion-Pair Chromatography]
CAS Number: 77-98-5
MDL Number: MFCD00009024
Molecular Formula: C8H25NO
Molecular Weight: 147.26
Form: Clear Liquid
Color: Colorless
Other Product Names / Synonyms: IPC-TEA-OH
Formula:
C₈H₂₁NO MW: 147.26 g/mol |
MDL Number:
MFCD00009024 CAS Number: 77-98-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...