Sodium dodecyl sulfate (SDS) ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Danger

I0352-500G I0352-25G I0352-100G
TCI0352-500GEA 166.7 USD
TCI0352-500G TCI0352-025G TCI0352-100G
Sodium dodecyl sulfate (SDS) ≥97.0% (by titrimetric analysis)
Sodium dodecyl sulphate (SDS)

[Reagent for Ion-Pair Chromatography] CAS Number: 151-21-3 MDL Number: MFCD00036175 Molecular Formula: C12H26O4S Molecular Weight: 288.38 Purity/Analysis Method: 97.0% (T) Form: Crystal Melting point (°C): 207 Flash Point (°C): 145


Product Categories: Analytical Science, HPLC Ion-Pair Reagents for Basic Samples Other Product Names / Synonyms: IPC-SDS, Laurylsulfuric Acid Sodium Salt, Dodecylsulfuric Acid Sodium Salt, Sodium Lauryl Sulfate

Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,II
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70133d1a576563b95828e0ed37231b08216ed2af&vid=0596d7502d61df67d5e9e3d5cf2392e080493266&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR