4-Fluorophenyl isocyanate 98,0

Supplier: TCI America

Synonyms: 4-Fluorophenylisocyanate, 1-Fluoro-4-isocyanatobenzene

I0339-25G I0339-5G
TCI0339-25GEA 152 USD
TCI0339-25G TCI0339-5G
4-Fluorophenyl isocyanate 98,0
4-Fluorophenyl isocyanate

CAS Number: 1195-45-5 MDL Number: MFCD00002023 Molecular Formula: C7H4FNO Molecular Weight: 137.11 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 68 Flash Point (°C): 53 Specific Gravity (20/20): 1.22 Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks Other Product Names / Synonyms: Isocyanic Acid 4-Fluorophenyl Ester

Formula: C₇H₄FNO
MW: 137.11 g/mol
Boiling Pt: 68 °C
Density: 1.22 (20 °C)
Flash Pt: 53 °C
MDL Number: MFCD00002023
CAS Number: 1195-45-5
UN: 3080
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7304f5cac5f89b82dd254e20f95719938a4b7b8&vid=b7304f5cac5f89b82dd254e20f95719938a4b7b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR