Imidazole ≥99.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine

I0290-25G
TCI0290-25GEA 156.9 USD
TCI0290-25G
Imidazole ≥99.0% (by GC, titration analysis)
Imidazole

[for Fluorimetric Analysis] CAS Number: 288-32-4 MDL Number: MFCD00005183 Molecular Formula: C3H4N2 Molecular Weight: 68.08 Purity/Analysis Method: 99.0% (GC,T) Form: Crystal Boiling point (°C): 256 Melting point (°C): 90 Flash Point (°C): 145

Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 256 °C
Melting Pt: 90 °C
Flash Pt: 145 °C
MDL Number: MFCD00005183
CAS Number: 288-32-4
UN: 2923
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f36d8bd46f6669875e9b14c372ee978e29510608&vid=1e331395861d6012f1a3a428540e4c4c088d6beb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR