Isoamyl alcohol ≥99.0%

Supplier: TCI America
Danger

I0289-500ML I0289-25ML
TCI0289-500MLEA 49.38 USD
TCI0289-500ML TCI0289-25ML
Isoamyl alcohol ≥99.0%
Isoamyl alcohol

CAS Number: 123-51-3 MDL Number: MFCD00002934 Molecular Formula: C5H12O Molecular Weight: 88.15 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 132 Flash Point (°C): 46 Specific Gravity (20/20): 0.81


Other Product Names / Synonyms: Isoamyl Alcohol, Isobutylcarbinol, Isopentanol, Isopentyl Alcohol

Formula: (CH₃)₂CHCH₂CH₂OH
Boiling Pt: 132 °C
Melting Pt: –117 °C
Density: 0.81 (20 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00002934
CAS Number: 123-51-3
EINECS: 204-633-5
UN: 1105
ADR: 3,III
Merck Index: 13,05214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f84b5fac2d9d87ddc43aef8ed62f60d80c864d8&vid=4f84b5fac2d9d87ddc43aef8ed62f60d80c864d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR