2-Propanol ≥99.5%

Supplier: TCI America
Danger

Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol

I0277-500ML
TCI0277-500MLEA 50.52 USD
TCI0277-500ML
2-Propanol ≥99.5%
2-Propanol

[for HPLC Solvent]
CAS Number: 67-63-0
MDL Number: MFCD00011674
Molecular Formula: C3H8O
Molecular Weight: 60.10
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Flash Point (°C): 15
Specific Gravity (20/20): 0.79


Product Categories: Analytical Science, Solvents for HPLC
Other Product Names / Synonyms: Isopropanol, 2-Propanol, IPA

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82 °C
Melting Pt: –89 °C
Density: 0.79 (20 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64f594c22f1639fcb8cd909677494ef2bc68a4ed&vid=c88b53c3a800ab998547247dfc17931fc3414614&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR