2,2,4-Trimethylpentane ≥99.0%

Supplier: TCI America
Danger

Synonyms: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

I0276-500ML
TCI0276-500MLEA 59.02 USD
TCI0276-500ML
2,2,4-Trimethylpentane ≥99.0%
2,2,4-Trimethylpentane

[for HPLC Solvent] CAS Number: 540-84-1 MDL Number: MFCD00008943 Molecular Formula: C8H18 Molecular Weight: 114.23 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 99 Melting point (°C): -107 Flash Point (°C): -9 Specific Gravity (20/20): 0.69


Product Categories: Analytical Science, Solvents for HPLC Other Product Names / Synonyms: Isooctane

Formula: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Boiling Pt: 99 °C
Melting Pt: –107 °C
Density: 0.69 (20 °C)
Flash Pt: –9 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3,II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cb8f538a543df8ef1e9a6d4eb14cac2e1055013&vid=3a73400337c36c617ce05c231d3cb9cd7928b7e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR