5-Iodo-2'-deoxyuridine (Idoxuridine) ≥99.0%

Supplier: TCI America

Synonyms: 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidine-2,4(1H,3H)-dione, Idoxuridine, (+)-5-Iodo-2'-deoxyuridine

I0258-1G I0258-25G I0258-5G
TCI0258-001GEA 45 USD
TCI0258-001G TCI0258-25G TCI0258-005G
5-Iodo-2'-deoxyuridine (Idoxuridine) ≥99.0%
5-Iodo-2'-deoxyuridine (Idoxuridine)

CAS Number: 54-42-2 MDL Number: MFCD00134656 Molecular Formula: C9H11IN2O5 Molecular Weight: 354.10 Purity/Analysis Method: 99.0% (T) Form: Crystal Specific rotation [a]20/D: 30 deg (C=1, 1mol/L NaOH sol.)


Product Categories: Bioscience, Antiviral Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs Other Product Names / Synonyms: Idoxuridine, 5-Iododesoxyuridine, IDU

Formula: C₉H₁₁IN₂O₅
MW: 354.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00134656
CAS Number: 54-42-2
EINECS: 200-207-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbf2b32a6a393135c6d12376b73b9ebb3a18a062&vid=bbf2b32a6a393135c6d12376b73b9ebb3a18a062&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR