Instant Buffer K40 pH 4.0 (acc. to Kolthoff)
Supplier: TCI America
I0234-1SET
TCI0234-SETEA
438.3
USD
TCI0234-SET
Instant Buffer K40 pH 4.0 (acc. to Kolthoff)
Buffers
pH Calibration Buffers
Form: White granules
Buffer prepared from a mixture of succinic acid and borax according to Kolthoff
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...