1-Naphthyl isothiocyanate ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 551-06-4
MDL Number: MFCD00003882
Molecular Formula: C11H7NS
Molecular Weight: 185.24
Purity/Analysis Method:
Product Categories: Bioscience, Reagents for Inflammation & Immunity Disorder Models, Analytical Science, UV Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: Isothiocyanic Acid 1-Naphthyl Ester
Formula:
C₁₁H₇NS Melting Pt: 55 °C |
MDL Number:
MFCD00003882 CAS Number: 551-06-4 UN: 2923 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...