Diisopropyl ether ≥99.0% stabilized

Supplier: TCI America
Danger

Synonyms: Isopropyl ether

I0172-500ML
TCI0172-500MLEA 46.51 USD
TCI0172-500ML
Diisopropyl ether ≥99.0% stabilized
Diisopropyl ether

(stabilized with BHT) CAS Number: 108-20-3 MDL Number: MFCD00008880 Molecular Formula: C6H14O Molecular Weight: 102.18 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 68 Melting point (°C): -60 Flash Point (°C): -28 Specific Gravity (20/20): 0.72


Other Product Names / Synonyms: Diisopropyl Ether

Formula: (CH₃)₂CHOCH(CH₃)₂
MW: 102.18 g/mol
Boiling Pt: 68 °C
Melting Pt: –60 °C
Density: 0.72 (20 °C)
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008880
CAS Number: 108-20-3
EINECS: 203-560-6
UN: 1159
ADR: 3,II
Merck Index: 13,05232
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=932b8c021b2bfa9a11b8d8812cafbafecc6f9239&vid=dea267e876203eb2c2674787701ec602da1b3cf6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR