Isonicotinaldehyde ≥96.0%

Supplier: TCI America

Synonyms: Pyridine-4-carboxaldehyde, 4-Pyridinecarboxaldehyde, 4-Pyridinecarbaldehyde

I0143-25ML
TCI0143-025MLEA 41.5 USD
TCI0143-025ML
Isonicotinaldehyde ≥96.0%
Isonicotinaldehyde

CAS Number: 872-85-5 MDL Number: MFCD00006425 Molecular Formula: C6H5NO Molecular Weight: 107.11 Purity/Analysis Method: 96.0% (GC) Form: Clear Liquid Boiling point (°C): 187 Flash Point (°C): 75 Specific Gravity (20/20): 1.14


Product Categories: Synthetic Organic Chemistry, Aldehydes (Multi-component Reaction) Other Product Names / Synonyms: Isonicotinaldehyde

Formula: C₆H₅NO
Boiling Pt: 187 °C
Density: 1.14 (20 °C)
Flash Pt: 75 °C
MDL Number: MFCD00006425
CAS Number: 872-85-5
EINECS: 212-832-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=218a790e03027da4fedf6ab49da0b57f26989aff&vid=218a790e03027da4fedf6ab49da0b57f26989aff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR