5-Iodoanthranilic acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Amino-5-iodobenzoic acid

I0049-5G I0049-25G
TCI0049-005GEA 31.4 USD
TCI0049-005G TCI0049-025G
5-Iodoanthranilic acid ≥98.0%
5-Iodoanthranilic acid

CAS Number: 5326-47-6 MDL Number: MFCD00007849 Molecular Formula: C7H6INO2 Molecular Weight: 263.03 Purity/Analysis Method: 98.0% (T) Form: Crystal


Other Product Names / Synonyms: 2-Amino-5-iodobenzoic Acid

Formula: C₇H₆INO₂
MW: 263.03 g/mol
Boiling Pt: 360
Melting Pt: 234
MDL Number: MFCD00007849
CAS Number: 5326-47-6
EINECS: 226-205-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea39f5223be0ddc353bcc8187fbac57cae24b273&vid=e65442dca8b0dc6edd39b46ece3c3f8b74755ba8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR