Invertose
Supplier: TCI America
Synonyms:
Invert sugar
I0043-25G
TCI0043-025GEA
115
USD
TCI0043-025G
Invertose
Invertose
CAS Number: 8013-17-0
MDL Number: MFCD00148911
Form: Crystal
Color: White
Specific rotation [a]20/D: -20 deg (C=5, H2O)
Product Categories: Bioscience, Sugars (Others), Basic Sugars (Mono & Oligosaccharides)
MDL Number:
MFCD00148911 CAS Number: 8013-17-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...