Indole ≥99.0%

Supplier: TCI America
Danger

Synonyms: 1H-Indole

I0021-500G I0021-25G I0021-100G
TCI0021-500GEA 240 USD
TCI0021-500G TCI0021-25G TCI0021-100G
Indole ≥99.0%
Indole

CAS Number: 120-72-9 MDL Number: MFCD00005607 Molecular Formula: C8H7N Molecular Weight: 117.15 Purity/Analysis Method: 99.0% (GC) Form: Crystal Boiling point (°C): 253 Melting point (°C): 53 Flash Point (°C): 122

Formula: C₈H₇N
MW: 117.15 g/mol
Boiling Pt: 253 °C
Melting Pt: 53 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 122 °C
MDL Number: MFCD00005607
CAS Number: 120-72-9
EINECS: 204-420-7
UN: 2811
ADR: 6.1 III
Merck Index: 13,04985
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d0985c6da19418f498fb4b4e83f5103cc828dbe&vid=5d0985c6da19418f498fb4b4e83f5103cc828dbe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR