Indene ≥93.0%
Supplier: TCI America
|
Danger
|
Synonyms:
1H-Indene
I0016-25ML
I0016-500ML
I0016-100ML
TCI0016-025MLEA
17.9
USD
TCI0016-025ML
TCI0016-500ML
TCI0016-100ML
Indene ≥93.0%
Indene
CAS Number: 95-13-6
MDL Number: MFCD00003777
Molecular Formula: C9H8
Molecular Weight: 116.16
Purity/Analysis Method:
Formula:
C₉H₈ Boiling Pt: 180 °C Melting Pt: –2 °C Density: 1.00 (20 °C) Flash Pt: 58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003777 CAS Number: 95-13-6 EINECS: 202-393-6 UN: 3295 ADR: 3,III Merck Index: 13,04964 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...