Indene ≥93.0%

Supplier: TCI America
Danger

Synonyms: 1H-Indene

I0016-25ML I0016-500ML I0016-100ML
TCI0016-025MLEA 17.9 USD
TCI0016-025ML TCI0016-500ML TCI0016-100ML
Indene ≥93.0%
Indene

CAS Number: 95-13-6 MDL Number: MFCD00003777 Molecular Formula: C9H8 Molecular Weight: 116.16 Purity/Analysis Method: 93.0% (GC) Form: Clear Liquid Boiling point (°C): 180 Flash Point (°C): 58 Freezing point (°C): -2 Specific Gravity (20/20): 1.00 Storage Temperature: 0°C

Formula: C₉H₈
Boiling Pt: 180 °C
Melting Pt: –2 °C
Density: 1.00 (20 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00003777
CAS Number: 95-13-6
EINECS: 202-393-6
UN: 3295
ADR: 3,III
Merck Index: 13,04964
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c3b5c4b4b6de51a3b4cdc2ef5bc29dadf42be77&vid=2c3b5c4b4b6de51a3b4cdc2ef5bc29dadf42be77&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR