Ninhydrin ≥98.0%

Supplier: TCI America
Warning

Synonyms: Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate

I0015-25G I0015-500G I0015-100G
TCI0015-025GEA 86.25 USD
TCI0015-025G TCI0015-500G TCI0015-100G
Ninhydrin ≥98.0%
Ninhydrin

CAS Number: 485-47-2 MDL Number: MFCD00003791 Molecular Formula: C9H6O4 Molecular Weight: 178.14 Purity/Analysis Method: 98.0% (T) Form: Crystal Melting point (°C): 250


Product Categories: Analytical Science, UV Detection (HPLC Labeling Reagents) Other Product Names / Synonyms: 2,2-Dihydroxy-1,3-indandione, 1,2,3-Indantrione Monohydrate, 1,2,3-Triketohydrindene Monohydrate

Formula: C₉H₆O₄
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 250 °C
Density: 0.862 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003791
CAS Number: 485-47-2
EINECS: 207-618-1
Merck Index: 13,06582
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f6a16695e76f490f3543afea5449d863358936d&vid=6f6a16695e76f490f3543afea5449d863358936d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR