Imidazole ≥99.8% (by GC), zone refined: number of passes 30

Supplier: TCI America

Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine

I0014-1SAMPLE
TCI0014-ZONEEA 141 USD
TCI0014-ZONE
Imidazole ≥99.8% (by GC), zone refined: number of passes 30
Imidazole

CAS Number: 288-32-4
MDL Number: MFCD00005183
Molecular Formula: C3H4N2
Molecular Weight: 68.08
Purity/Analysis Method: >99.8% (GC)
Form: Crystal
Melting point (°C): 92
Flash Point (°C): 145

Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 255
Melting Pt: 92 °C
Flash Pt: 145 °C
MDL Number: MFCD00005183
CAS Number: 288-32-4
UN: 2923
ADR: 8 III
Merck Index: 13,04935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=483a0fe09dfd0580c10d4e2b45d6ccb16d4fff6d&vid=26f0b2de846747291fb144df2c567c4795c7b974&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.8% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR