Imidazole ≥98.0%

Supplier: TCI America
Danger

Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine

I0001-500G I0001-25G I0001-100G
TCI0001-500GEA 139.11 USD
TCI0001-500G TCI0001-25G TCI0001-100G
Imidazole ≥98.0%
Imidazole

CAS Number: 288-32-4 MDL Number: MFCD00005183 Molecular Formula: C3H4N2 Molecular Weight: 68.08 Purity/Analysis Method: 98.0% (T) Form: Crystal Boiling point (°C): 256 Melting point (°C): 90 Flash Point (°C): 145


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents, Bioscience, Reagents for Oligosaccharide Synthesis Other Product Names / Synonyms: Glyoxaline

Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 255
Melting Pt: 90 °C
Density: 1.18 g/cm³ (20 °C)
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 2923
ADR: 8 III
Merck Index: 13,04935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d27c5226abf7468e9bec014d556f5fb54ad6b5eb&vid=d27c5226abf7468e9bec014d556f5fb54ad6b5eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR