Hydroxylammonium chloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Hydroxylamine HCl, Hydroxylamine hydrochloride

H1581-25G H1581-500G
TCH1581-25GEA 16.04 USD
TCH1581-25G TCH1581-500G
Hydroxylammonium chloride ≥97.0% (by titrimetric analysis)
Hydroxylammonium chloride
Formula: NH₂OH·HCl
MW: 69.49 g/mol
Boiling Pt: 306
Melting Pt: 151
MDL Number: MFCD00051089
CAS Number: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8 III
Merck Index: 14,04828

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Redox method) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR