Hexafluorocyclotriphosphazene ≥97.0% (by GC)
Supplier: TCI America
H1429-25G
H1429-5G
TCH1429-25GEA
608
USD
TCH1429-25G
TCH1429-5G
Hexafluorocyclotriphosphazene ≥97.0% (by GC)
Hexafluorocyclotriphosphazene
CAS Number: 15599-91-4
MDL Number: MFCD00192400
Molecular Formula: F6N3P3
Molecular Weight: 248.93
Purity/Analysis Method:
Formula:
F₆N₃P₃ Boiling Pt: 51 °C Melting Pt: 27 °C |
MDL Number:
MFCD00192400 CAS Number: 15599-91-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...