4-Hydroxy-6-methylcoumarin ≥98.0% (by GC)

Supplier: TCI America

H1421-1G H1421-5G
TCH1421-1GEA 31 USD
TCH1421-1G TCH1421-5G
4-Hydroxy-6-methylcoumarin ≥98.0% (by GC)
4-Hydroxy-6-methylcoumarin

CAS Number: 13252-83-0 MDL Number: MFCD00085738 Molecular Formula: C10H8O3 Molecular Weight: 176.17 Purity/Analysis Method: 98.0% (GC) Form: Crystal Melting point (°C): 261 Lambda max.: 309 nm (EtOH aq.)

Formula: C₁₀H₈O₃
Melting Pt: 261 °C
MDL Number: MFCD00085738
CAS Number: 13252-83-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=296656aa9fa30eafb9af506760c612a340783249&vid=296656aa9fa30eafb9af506760c612a340783249&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR