4-Hydroxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
(4-Hydroxyphenyl)metaboric acid, Dihydroxy(4-hydroxyphenyl)borane, 4-Hydroxybenzeneboronic acid
H1228-1G
H1228-5G
TCH1228-1GEA
44.77
USD
TCH1228-1G
TCH1228-5G
4-Hydroxyphenylboronic acid (contains varying amounts of Anhydride)
4-Hydroxyphenylboronic acid
CAS Number: 71597-85-8
MDL Number: MFCD01074628
Molecular Formula: C6H7BO3
Molecular Weight: 137.93
Form: Crystal
Color: White
Melting point (°C): 232
Other Product Names / Synonyms: 4-Hydroxybenzeneboronic Acid
Formula:
C₆H₇BO₃ MW: 137.93 g/mol Melting Pt: 232 °C |
MDL Number:
MFCD01074628 CAS Number: 71597-85-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...