Hydrogen chloride ∼4 mol/L in 1,4-dioxane

Supplier: TCI America

H1062-100ML
TCH1062-25MLEA 56.83 USD
TCH1062-25ML TCH1062-500ML TCH1062-100ML
Hydrogen chloride ∼4 mol/L in 1,4-dioxane
Hydrogen chloride

CAS Number: 7647-01-0
MDL Number: MFCD00011324
Molecular Formula: HCl
Molecular Weight: 36.46
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 17

Formula: HCl
Flash Pt: 17 °C
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1050
ADR: 2,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR