(1-Hexadecyl)tributylphosphonium bromide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
H1047-25G
TCH1047-25GEA
148.56
USD
TCH1047-25G
(1-Hexadecyl)tributylphosphonium bromide ≥98.0% (by titrimetric analysis)
(1-Hexadecyl)tributylphosphonium bromide
CAS Number: 14937-45-2
MDL Number: MFCD00011775
Molecular Formula: C28H60BrP
Molecular Weight: 507.67
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Ionic Liquids
Other Product Names / Synonyms: Cetyltributylphosphonium Bromide, Hexadecyltributylphosphonium Bromide
Formula:
C₂₈H₆₀BrP Melting Pt: 61 °C |
MDL Number:
MFCD00011775 CAS Number: 14937-45-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...