4'-Hydroxychalcone ≥95.0%
Supplier: TCI America
Synonyms:
1-(4-Hydroxyphenyl)-3-phenylprop-2-en-1-one, 4'-Hydroxybenzylidenacetophenone
H0945-5G
TCH0945-005GEA
23
USD
TCH0945-005G
4'-Hydroxychalcone ≥95.0%
4'-Hydroxychalcone
CAS Number: 2657-25-2
MDL Number: MFCD00016484
Molecular Formula: C15H12O2
Molecular Weight: 224.26
Purity/Analysis Method:
Product Categories: Bioscience, Aromatic Polyketides
Other Product Names / Synonyms: 2-Benzal-4'-hydroxyacetophenone, 2-Benzylidene-4'-hydroxyacetophenone
Formula:
C₁₅H₁₂O₂ MW: 224.26 g/mol Melting Pt: 174 °C |
MDL Number:
MFCD00016484 CAS Number: 2657-25-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...