(Hydrazinocarbonyl)ferrocene ≥95.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Ferrocenecarbohydrazide
H0941-1G
TCH0941-001GEA
405.9
USD
TCH0941-001G
(Hydrazinocarbonyl)ferrocene ≥95.0% (by HPLC, titration analysis)
(Hydrazinocarbonyl)ferrocene
[for HPLC Labeling]
CAS Number: 12153-28-5
MDL Number: MFCD01321175
Molecular Formula: C11H12FeN2O
Molecular Weight: 244.08
Purity/Analysis Method:
Product Categories: Analytical Science, Electrochemical Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: Ferrocenecarbohydrazide
Formula:
C₁₁H₁₂FeN₂O MW: 244.08 g/mol |
MDL Number:
MFCD01321175 CAS Number: 12153-28-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...