4-Hydroxy-3,5-dimethylbenzoic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
H0938-5G
TCH0938-005GEA
114
USD
TCH0938-005G
4-Hydroxy-3,5-dimethylbenzoic acid ≥98.0% (by titrimetric analysis)
4-Hydroxy-3,5-dimethylbenzoic acid
CAS Number: 4919-37-3
MDL Number: MFCD00016536
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method:
Formula:
C₉H₁₀O₃ Melting Pt: 225 °C |
MDL Number:
MFCD00016536 CAS Number: 4919-37-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...