Lithiumbis(trimethylsilyl)amide ∼26% ∼1.3 mol/L in THF

Supplier: TCI America

Synonyms: Lithiumbis(triméthylsilyl)amide, HMDS-Li, N-Lithiohexamethyldisilazane, HMDS lithium salt

H0915-500ML
TCH0915-500MLEA 282.85 USD
TCH0915-500ML
Lithiumbis(trimethylsilyl)amide ∼26% ∼1.3 mol/L in THF
Lithium bis(trimethylsilyl)amide

CAS Number: 4039-32-1
MDL Number: MFCD00008261
Molecular Formula: C6H18LiNSi2
Molecular Weight: 167.33
Form: Clear Liquid
Color: Yellow
Flash Point (°C): -17


Other Product Names / Synonyms: Hexamethyldisilazane Lithium Salt, LiHMDS, Lithium Hexamethyldisilazide, LHMDS

Formula: C₆H₁₈LiNSi₂
MW: 167.33 g/mol
Flash Pt: –17 °C
MDL Number: MFCD00008261
CAS Number: 4039-32-1
UN: 3396
ADR: 4.3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50687490dc17fc6d3552cd95830fa707d47bedd2&vid=eb5e8ba8c5b64ae56c2e37f26dc51cfc40b65a75&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR