3-Hydroxyglutaronitrile ≥95.0%

Supplier: TCI America

H0897-5G
TCH0897-005GEA 343.1 USD
TCH0897-005G
3-Hydroxyglutaronitrile ≥95.0%
3-Hydroxyglutaronitrile

CAS Number: 13880-89-2 MDL Number: MFCD00671545 Molecular Formula: C5H6N2O Molecular Weight: 110.12 Purity/Analysis Method: 95.0% (GC) Form: Clear Liquid Boiling point (°C): 203 Specific Gravity (20/20): 1.14 Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1,3-Dicyano-2-propanol

Formula: C₅H₆N₂O
Boiling Pt: 203 °C
Density: 1.14 (20 °C)
MDL Number: MFCD00671545
CAS Number: 13880-89-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77b38d1ed734d097887af81746400af35a435857&vid=77b38d1ed734d097887af81746400af35a435857&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR