Methyl-5-hydroxynicotinate ≥98.0%
Supplier: TCI America
Synonyms:
Methyl 5-hydroxynicotinate, Methyl 5-hydroxy-3-pyridinecarboxylate
H0841-25G
H0841-5G
H0841-1G
TCH0841-25GEA
728.1
USD
TCH0841-25G
TCH0841-005G
TCH0841-001G
Methyl-5-hydroxynicotinate ≥98.0%
Methyl 5-hydroxynicotinate
CAS Number: 30766-22-4
MDL Number: MFCD00191521
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method:
Other Product Names / Synonyms: 5-Hydroxynicotinic Acid Methyl Ester, 5-Hydroxy-3-pyridinecarboxylic Acid Methyl Ester, Methyl 5-Hydroxy-3-pyridinecarboxylate
Formula:
C₇H₇NO₃ Melting Pt: 190 °C |
MDL Number:
MFCD00191521 CAS Number: 30766-22-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...