Methyl-5-hydroxynicotinate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 5-hydroxynicotinate, Methyl 5-hydroxy-3-pyridinecarboxylate

H0841-25G H0841-5G H0841-1G
TCH0841-25GEA 728.1 USD
TCH0841-25G TCH0841-005G TCH0841-001G
Methyl-5-hydroxynicotinate ≥98.0%
Methyl 5-hydroxynicotinate

CAS Number: 30766-22-4 MDL Number: MFCD00191521 Molecular Formula: C7H7NO3 Molecular Weight: 153.14 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Melting point (°C): 190


Other Product Names / Synonyms: 5-Hydroxynicotinic Acid Methyl Ester, 5-Hydroxy-3-pyridinecarboxylic Acid Methyl Ester, Methyl 5-Hydroxy-3-pyridinecarboxylate

Formula: C₇H₇NO₃
Melting Pt: 190 °C
MDL Number: MFCD00191521
CAS Number: 30766-22-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1036875bbc12d524975b5f0972c328ffb2da7845&vid=1036875bbc12d524975b5f0972c328ffb2da7845&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR