4-Hydroxythiophenol ≥97.0% (by GC, titration analysis)

Supplier: TCI America

H0662-25G
TCH0662-025GEA 150.55 USD
TCH0662-025G
4-Hydroxythiophenol ≥97.0% (by GC, titration analysis)
4-Hydroxythiophenol

CAS Number: 637-89-8 MDL Number: MFCD00004850 Molecular Formula: C6H6OS Molecular Weight: 126.17 Purity/Analysis Method: 97.0% (GC,T) Form: Crystal Flash Point (°C): 110 Freezing point (°C): 34


Other Product Names / Synonyms: 4-Hydroxythiophenol, 4-Mercaptophenol

Formula: C₆H₆OS
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: 33…35 °C
Flash Pt: 110 °C
MDL Number: MFCD00004850
CAS Number: 637-89-8
EINECS: 211-307-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c1179c561b4c261fb7ac4fcb0d41bbccc32c861&vid=4c1179c561b4c261fb7ac4fcb0d41bbccc32c861&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR