5-Hydroxy-1-tetralone ≥99.0%

Supplier: TCI America

Synonyms: 5-Hydroxy-3,4-dihydronaphthalen-1(2H)-one, 5-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one

H0614-1G H0614-25G H0614-5G
TCH0614-1GEA 36 USD
TCH0614-1G TCH0614-25G TCH0614-5G
5-Hydroxy-1-tetralone ≥99.0%
5-Hydroxy-1-tetralone

CAS Number: 28315-93-7 MDL Number: MFCD00001693 Molecular Formula: C10H10O2 Molecular Weight: 162.19 Purity/Analysis Method: 99.0% (GC) Form: Crystal Melting point (°C): 208


Other Product Names / Synonyms: 5-Hydroxy-3,4-dihydronaphthalen-1(2H)-one

Formula: C₁₀H₁₀O₂
Melting Pt: 208 °C
MDL Number: MFCD00001693
CAS Number: 28315-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f997c4c257869b41f2825f1f01293edd870e14b4&vid=f997c4c257869b41f2825f1f01293edd870e14b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR