5-Hydroxy-1-tetralone ≥99.0%
Supplier: TCI America
Synonyms:
5-Hydroxy-3,4-dihydronaphthalen-1(2H)-one, 5-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one
H0614-1G
H0614-25G
H0614-5G
TCH0614-1GEA
36
USD
TCH0614-1G
TCH0614-25G
TCH0614-5G
5-Hydroxy-1-tetralone ≥99.0%
5-Hydroxy-1-tetralone
CAS Number: 28315-93-7
MDL Number: MFCD00001693
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method:
Other Product Names / Synonyms: 5-Hydroxy-3,4-dihydronaphthalen-1(2H)-one
Formula:
C₁₀H₁₀O₂ Melting Pt: 208 °C |
MDL Number:
MFCD00001693 CAS Number: 28315-93-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...