4-Biphenylylmethanol ≥99.0%
Supplier: TCI America
CAS Number: 3597-91-9
MDL Number: MFCD00004660
Molecular Formula: C13H12O
Molecular Weight: 184.24
Purity/Analysis Method:
Other Product Names / Synonyms: Biphenyl-4-methanol, 4-Phenylbenzyl Alcohol
Formula:
C₁₃H₁₂O Boiling Pt: 184 °C Melting Pt: 104 °C |
MDL Number:
MFCD00004660 CAS Number: 3597-91-9 EINECS: 222-745-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...