2,2,3,3,4,4,4-Heptafluoro-1-butanol ≥95.0%
Supplier: TCI America
|
Danger
|
Synonyms:
1H,1H-Perfluoro-1-butanol, 2,2,3,3,4,4,4-Heptafluorobutan-1-ol, Heptafluoro-1-butanol
H0548-5G
H0548-25G
TCH0548-005GEA
57
USD
TCH0548-005G
TCH0548-025G
2,2,3,3,4,4,4-Heptafluoro-1-butanol ≥95.0%
2,2,3,3,4,4,4-Heptafluoro-1-butanol
CAS Number: 375-01-9
MDL Number: MFCD00004674
Molecular Formula: C4H3F7O
Molecular Weight: 200.06
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks, Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1H,1H-Heptafluoro-1-butanol
Formula:
C₄H₃F₇O MW: 200.06 g/mol Boiling Pt: 96 Density: 1.6 g/cm³ (20 °C) Flash Pt: 25 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004674 CAS Number: 375-01-9 EINECS: 206-782-1 UN: 1987 ADR: 3 III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...