11ɑ-Hydroxyprogesterone ≥95.0%
Supplier: TCI America
Synonyms:
11ɑ-Hydroxypregn-4-ene-3,20-dione
H0498-1G
TCH0498-001GEA
207.1
USD
TCH0498-001G
11ɑ-Hydroxyprogesterone ≥95.0%
11alpha-Hydroxyprogesterone
CAS Number: 80-75-1
MDL Number: MFCD00003652
Molecular Formula: C21H30O3
Molecular Weight: 330.47
Purity/Analysis Method:
Product Categories: Bioscience, Pregnanes (Progestogen), Steroids
Other Product Names / Synonyms: 11alpha-Hydroxypregn-4-ene-3,20-dione
Formula:
C₂₁H₃₀O₃ MW: 330.47 g/mol Melting Pt: 169 °C |
MDL Number:
MFCD00003652 CAS Number: 80-75-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...