n-Hexane ≥96.0%

Supplier: TCI America
Danger

H0490-500ML
TCH0490-500MLEA 101 USD
TCH0490-500ML
n-Hexane ≥96.0%
n-Hexane

[for HPLC Solvent]
CAS Number: 110-54-3
MDL Number: MFCD00009520
Molecular Formula: C6H14
Molecular Weight: 86.18
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 68
Melting point (°C): -95
Flash Point (°C): -22
Specific Gravity (20/20): 0.66

Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 68 °C
Melting Pt: –95 °C
Density: 0.66 (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a28017b9745da0fad27d22a5c53709a10aff7d0&vid=5d8dabbca4150d2eb34fe0caa1eb73b57ca61978&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR