1-(2-Hydroxyethyl)pyrrolidine ≥98.0%

Supplier: TCI America

Synonyms: 1-Pyrrolidineethanol, N-(2-Hydroxyethyl)pyrrolidine

H0479-25ML
TCH0479-025MLEA 51 USD
TCH0479-025ML
1-(2-Hydroxyethyl)pyrrolidine ≥98.0%
1-(2-Hydroxyethyl)pyrrolidine

CAS Number: 2955-88-6 MDL Number: MFCD00003181 Molecular Formula: C6H13NO Molecular Weight: 115.18 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 187 Flash Point (°C): 56 Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: 1-Pyrrolidineethanol

Formula: C₆H₁₃NO
Boiling Pt: 187 °C
Density: 0.98 (20 °C)
Flash Pt: 56 °C
MDL Number: MFCD00003181
CAS Number: 2955-88-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05594c51e071b46b255a6b5599afa34bbd5b280d&vid=05594c51e071b46b255a6b5599afa34bbd5b280d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR