1-Hexanesulfonic acid sodium salt ≥98.0%
Supplier: TCI America
Synonyms:
Sodium hexane-1-sulfonate, Sodium hexane-1-sulphonate, Sodium hexanesulfonate, Sodium hexanesulphonate, Sodium 1-hexanesulfonate, Sodium 1-hexanesulphonate
H0447-25G
H0447-500G
H0447-100G
TCH0447-25GEA
100.4
USD
TCH0447-25G
TCH0447-500G
TCH0447-100G
1-Hexanesulfonic acid sodium salt ≥98.0%
1-Hexanesulphonic acid sodium salt
CAS Number: 2832-45-3
MDL Number: MFCD00007542
Molecular Formula: C6H14O3S
Molecular Weight: 188.22
Purity/Analysis Method:
Other Product Names / Synonyms: 1-Hexanesulfonic Acid Sodium Salt, Hexylsulfonic Acid Sodium Salt
Formula:
C₆H₁₃O₃SNa MW: 188.22 g/mol Melting Pt: 300 Storage Temperature: Ambient |
MDL Number:
MFCD00007542 CAS Number: 2832-45-3 EINECS: 220-601-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...