1,1,1,3,3,3-Hexafluoro-2-propanol ≥99.0%

Supplier: TCI America
Danger

H0424-25G
TCH0424-25GEA 92.36 USD
TCH0424-25G
1,1,1,3,3,3-Hexafluoro-2-propanol ≥99.0%
1,1,1,3,3,3-Hexafluoro-2-propanol

CAS Number: 920-66-1 MDL Number: MFCD00011651 Molecular Formula: C3H2F6O Molecular Weight: 168.04 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 58 Melting point (°C): -4 Specific Gravity (20/20): 1.62 Lambda max.: 229 nm


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks Other Product Names / Synonyms: Hexafluoroisopropyl Alcohol, HFIP, HFP

Formula: C₃H₂F₆O
Boiling Pt: 58 °C
Melting Pt: –4 °C
Density: 1.62 (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011651
CAS Number: 920-66-1
EINECS: 213-059-4
UN: 1760
ADR: 8,II
Merck Index: 13,09598
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e234f1e3f6f22316f972e1aa8e04d2033f5b73e5&vid=e234f1e3f6f22316f972e1aa8e04d2033f5b73e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR