1-Hexacosanol ≥95.0%

Supplier: TCI America

Synonyms: 1-Hydroxy hexacosane, 1-Hexacosyl alcohol, Cerotyl alcohol, Pentacosyl carbinol

H0342-100MG H0342-1G
TCH0342-100MGEA 174.09 USD
TCH0342-100MG TCH0342-001G
1-Hexacosanol ≥95.0%
1-Hexacosanol

CAS Number: 506-52-5 MDL Number: MFCD00002940 Molecular Formula: C26H54O Molecular Weight: 382.72 Purity/Analysis Method: 95.0% (GC) Form: Crystal Melting point (°C): 80


Product Categories: Bioscience, Higher Fatty Acids & Higher Alcohols Other Product Names / Synonyms: Ceryl Alcohol

Formula: H₃C(CH₂)₂₄CH₂OH
MW: 382.71 g/mol
Boiling Pt: 412
Melting Pt: 80 °C
Density: 0.8215 g/cm³ (85 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002940
CAS Number: 506-52-5
EINECS: 208-044-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a0f95437c5a5a40980cbf5156c242146fe29229&vid=2bf521caad1e9abfd34b388ebe58aaa3fc16661a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR